Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HLCL-61 hydrochloride | 1158279-20-9 | MFCD09446681 | 99.9% | 380.91 g/mol | C23H25ClN2O | 1 ML
SDP

Catalog No. 5000215424
Encompass_Preferred

HLCL-61 hydrochloride is a small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5) supplied for research use. It is provided as concentrated DMSO stock solutions and as solid material for in vitro and in vivo studies; not for human or veterinary use.

  • First-in-class PRMT5 inhibitor for research applications.
  • Available as 10 mM stock solutions in DMSO and as solids in multiple sizes.
  • High reported purity (approximately 99.9%).
  • Soluble in DMSO (≈131 mM at 50 mg/mL) and poorly soluble in water.
  • Storage: sealed at 4°C for solids; in solvent -80°C for long-term storage.
  • Identifying information: CAS number 1158279-20-9; molecular weight ≈380.91 g/mol.

Catalog No. 50-002-15424 Supplier Medchemexpress LLC Supplier No. HY100025A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HLCL-61 hydrochloride is a small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5) supplied for research use. It is provided as concentrated DMSO stock solutions and as solid material for in vitro and in vivo studies; not for human or veterinary use.

  • First-in-class PRMT5 inhibitor for research applications.
  • Available as 10 mM stock solutions in DMSO and as solids in multiple sizes.
  • High reported purity (approximately 99.9%).
  • Soluble in DMSO (≈131 mM at 50 mg/mL) and poorly soluble in water.
  • Storage: sealed at 4°C for solids; in solvent -80°C for long-term storage.
  • Identifying information: CAS number 1158279-20-9; molecular weight ≈380.91 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.