Learn More
Medchemexpress LLC HLCL-61 hydrochloride | 1158279-20-9 | MFCD09446681 | 99.9% | 380.91 g/mol | C23H25ClN2O | 25 MG
SDP
Catalog No. 5000215086
Encompass_Preferred
HLCL-61 hydrochloride is a research-grade small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5), supplied as a hydrochloride salt for laboratory use only. It is intended for in vitro and preclinical research and is not for human or veterinary use.
  • Inhibits PRMT5 activity in biochemical and cellular assays.
  • High reported purity of 99.87%.
  • Molecular weight is 380.91 g/mol.
  • Identified by CAS number 1158279-20-9 for unambiguous chemical identification.
  • Available in milligram quantities suitable for laboratory studies, including a 25 mg pack size.
Catalog No. 50-002-15086 Supplier Medchemexpress LLC Supplier No. HY100025A25MG
HLCL-61 hydrochloride is a research-grade small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5), supplied as a hydrochloride salt for laboratory use only. It is intended for in vitro and preclinical research and is not for human or veterinary use.
  • Inhibits PRMT5 activity in biochemical and cellular assays.
  • High reported purity of 99.87%.
  • Molecular weight is 380.91 g/mol.
  • Identified by CAS number 1158279-20-9 for unambiguous chemical identification.
  • Available in milligram quantities suitable for laboratory studies, including a 25 mg pack size.