Learn More
Medchemexpress LLC HLCL-61 hydrochloride | 1158279-20-9 | MFCD09446681 | 99.9% | 380.91 g/mol | C23H25ClN2O | 50 MG
SDP
Catalog No. 5000215512
Encompass_Preferred
HLCL-61 hydrochloride is a first-in-class small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5), provided as a solid research reagent with characterized purity and storage recommendations for biochemical and cellular studies.
  • First-in-class PRMT5 inhibitor for research applications.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a solid, white to off-white powder.
  • Documented molecular formula and molecular weight for experiment planning.
  • Storage guidance provided for solvent and dry conditions to preserve stability.
  • For research use only; not for human or animal therapeutic use.
Catalog No. 50-002-15512 Supplier Medchemexpress LLC Supplier No. HY100025A50MG
HLCL-61 hydrochloride is a first-in-class small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5), provided as a solid research reagent with characterized purity and storage recommendations for biochemical and cellular studies.
  • First-in-class PRMT5 inhibitor for research applications.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a solid, white to off-white powder.
  • Documented molecular formula and molecular weight for experiment planning.
  • Storage guidance provided for solvent and dry conditions to preserve stability.
  • For research use only; not for human or animal therapeutic use.