Learn More
TARGETMOL CHEMICALS INC HLCL-61 HYDROCHLORIDE 25MG
SDP
Catalog No. 502576432
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HLCL-61 hydrochloride (HLCL-61 HCL) is a potent and selective PRMT5 inhibitor for the treatment of acute myeloid leukemia. purity: 99%
Catalog No. 50-257-6432 Supplier TARGETMOL CHEMICALS INC Supplier No. T685725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HLCL-61 hydrochloride (HLCL-61 HCL) is a potent and selective PRMT5 inhibitor for the treatment of acute myeloid leukemia. purity: 99%