Learn More
Cayman Chemical HLCL-61 (hydrochloride), Purity: ≥98%, CAS: 1158279-20-9, 10 mg
SDP
Catalog No. 502984599
Encompass_Preferred
A PRMT5 inhibitor; inhibits the growth of multiple AML cell lines and patient-derived tumor samples (IC50s = 7.21-21.46 and 3.98-8.72 μM, respectively); selective for PRMT5, lacking activity against PRMT1, PRMT4, and PRMT7 in an enzyme assay; induces myeloid differentiation of THP-1 cells and increases CD11b expression in a dose-dependent manner; increases expression of miR-29b mRNA resulting in a 4-fold decrease in FLT3 activity in THP-1 cells expressing an FLT3 luciferase reporter
Catalog No. 50-298-4599 Supplier Cayman Chemical Supplier No. 1989710 MG
A PRMT5 inhibitor; inhibits the growth of multiple AML cell lines and patient-derived tumor samples (IC50s = 7.21-21.46 and 3.98-8.72 μM, respectively); selective for PRMT5, lacking activity against PRMT1, PRMT4, and PRMT7 in an enzyme assay; induces myeloid differentiation of THP-1 cells and increases CD11b expression in a dose-dependent manner; increases expression of miR-29b mRNA resulting in a 4-fold decrease in FLT3 activity in THP-1 cells expressing an FLT3 luciferase reporter