Learn More
Aviva Systems Biology Corp HLTF Antibody - N-terminal region (ARP38217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLTF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HLTF. Alternate names: HIP116, HIP116A, HLTF1, RNF80, SMARCA3, SNF2L3, ZBU1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-56568