Learn More
Electron Microscopy Sciences Holey Carbon Film with Carbon/Formvar 1 nm / 5 nm thick on 200 mesh Gold Grid
SDP
Catalog No. 501931396
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Gold
Catalog No. 50-193-1396 Supplier Electron Microscopy Sciences Supplier No. HC200AUFCF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Gold