Learn More
Electron Microscopy Sciences Holey Carbon Film with Carbon/Formvar 1 nm / 5 nm thick on 200 mesh Nickel Grid
SDP
Catalog No. 501931393
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Nickel
Catalog No. 50-193-1393 Supplier Electron Microscopy Sciences Supplier No. HC200NIFCF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Nickel