Learn More
Electron Microscopy Sciences Holey Carbon Film with Carbon/Formvar 1 nm / 5 nm thick on 400 mesh Nickel Grid
SDP
Catalog No. 501931395
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Nickel
Catalog No. 50-193-1395 Supplier Electron Microscopy Sciences Supplier No. HC400NIFCF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon 1 nm, Formvar 5 nm. Supported by a mesh grid.
Grid: Nickel