Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Carbon < 3 nm thick on 200 mesh Gold Grid
SDP
Catalog No. 501931378
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Gold
Catalog No. 50-193-1378 Supplier Electron Microscopy Sciences Supplier No. HC200AUCC
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Gold