Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Carbon < 3 nm thick on 300 mesh Nickel Grid
SDP
Catalog No. 501931376
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Nickel
Catalog No. 50-193-1376 Supplier Electron Microscopy Sciences Supplier No. HC300NICC
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Carbon < 3 nm. Supported by a mesh grid.
Grid: Nickel