Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Formvar 3-4 nm thick on 200 mesh Copper Grid
SDP
Catalog No. 501931381
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Copper
Catalog No. 50-193-1381 Supplier Electron Microscopy Sciences Supplier No. HC200CUFF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Copper