Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Formvar 3-4 nm thick on 200 mesh Nickel Grid
SDP
Catalog No. 501931384
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Nickel
Catalog No. 50-193-1384 Supplier Electron Microscopy Sciences Supplier No. HC200NIFF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Nickel