Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Formvar 3-4 nm thick on 300 mesh Gold Grid
SDP
Catalog No. 501931388
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Gold
Catalog No. 50-193-1388 Supplier Electron Microscopy Sciences Supplier No. HC300AUFF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Gold