Learn More
Electron Microscopy Sciences Holey Carbon Film with Ultrathin Formvar 3-4 nm thick on 400 mesh Copper Grid
SDP
Catalog No. 501931383
Encompass_Preferred
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Copper
Catalog No. 50-193-1383 Supplier Electron Microscopy Sciences Supplier No. HC400CUFF
The continuous ultrathin film on holey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. Thickness: Formvar 3-4 nm. Supported by a mesh grid.
Grid: Copper