Learn More
Medchemexpress LLC HY-122816 5mg Medchemexpress, HLY78 CAS:854847-61-3 Purity:>98%
SDP
Catalog No. 501932572
Encompass_Preferred
Medchemexpress, HY-122816 5mg HLY78 CAS:854847-61-3 HLY78 is an activator of the Wnt/β-catenin signaling pathway, which targets the DIX domain of Axin and potentiates the Axin-LRP6 association to promote Wnt signaling transduction [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2572 Supplier Medchemexpress LLC Supplier No. HY1228165MG
Medchemexpress, HY-122816 5mg HLY78 CAS:854847-61-3 HLY78 is an activator of the Wnt/β-catenin signaling pathway, which targets the DIX domain of Axin and potentiates the Axin-LRP6 association to promote Wnt signaling transduction [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.