Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories

Catalog No. 1407136 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Blade Holder, hi-profile blade
Blade Holder, lo-profile blade
Brain Rsrch Freezing Platform
Height Adjustment Plate, 10 mm
Height Adjustment Plate, 20 mm
Height Adjustment Plate, 5 mm
Std Specimen Clamp w/ Tray
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-071-36 Height Adjustment Plate, 10 mm
14-071-37 Height Adjustment Plate, 20 mm
14-070-85 Height Adjustment Plate, 5 mm
70-520-0 Blade Holder, lo-profile blade
14-071-07 Blade Holder, hi-profile blade
715480 Std Specimen Clamp w/ Tray
98-100-0 Brain Rsrch Freezing Platform
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-071-36 Supplier Epredia™ Supplier No. 440400
Add to Cart
Add to Cart

Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
  • Cooling element for buffer solution

  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
  • Large-field magnifier

  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
  • StemiScope™ with optics and mount

  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
  • Swan-neck light guide

  • 3.5 mm fiber bundle
  • Swan-neck light guide: 50 mm long
  • Flexible light guide: 750 mm long
  • High light intensity
  • Include adapter for the KL200 (760317)
  • Specifications

    Product Type Height Adjustment Plate, 10 mm
    For Use With (Equipment) Epredia HM 430 and HM 450 Sliding Microtomes
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.