Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HM 650V Vibrating Blade Microtome Specimen Stage F (Flat)for medium to large specimens or tissue (from 10mm to 45mm)

Catalog No. 152530 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Adapter R
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-253-0 Adapter R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-253-0 Supplier Epredia™ Supplier No. 152530
Only null left
Add to Cart
Add to Cart

Use these Epredia™ HM 650V Vibrating Blade Microtome Accessories with the Epredia™ HM650V Vibrating Blade Microtome.

Standard buffer tray:
  • Hold stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution:
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier:
  • Magnification: 2.5x
  • Ability to move, swivel, and displace for optimal viewing position
  • Adapter and illumination included
StemiScope™ with optics and mount:
  • Eyepieces: 10x
  • Supplementary lens: 0.63x
  • Zoom magnifying range: 4.1 to 31.5x
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide:
  • 3.5mm fiber bundle
  • 50mm long swan-neck light guide
  • 750mm long flexible light guide
  • High light intensity
  • Adapter for the KL200 (760317) included

Specifications

Product Type Adapter R
For Use With (Equipment) Razor Blades
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.