Learn More
Gilson Company Inc HM-70A
Supplier:  Gilson Company Inc HM70A
Encompass_Preferred
  • Robust and automated instrument for determining aggregate degradation resistance.
  • Measures abrasion and impact resistance according to ASTM and AASHTO standards.
  • Features electronic interlock for enhanced operator safety.
  • Sound-attenuating enclosure reduces operational noise for a comfortable laboratory environment.
  • Ergonomic design with user-friendly external controls.
  • 1 hp motor ensures precise drum rotation and efficient sample loading.
  • Drum rotation speed ranges from 30 to 33 rpm.
  • Drum made from 1/2 in thick welded steel for durability.
  • 28 in drum diameter accommodates various aggregate sizes.
  • Includes L.A. Abrasion Machine, catch pan, and twelve 46-48 mm hardened steel balls.
  • Dimensions: 38 x 40 x 46.5 in.
  • Uncrated weight: Approximately 1,050 lbs.
Catalog No. NC3590282
Only null left
Add to Cart
Request a Quote