Learn More
MEDNA SCIENTIFIC INC Human IgG1 Poly-Specificity Control Antibody - 1mg
Supplier:  MEDNA SCIENTIFIC INC H1311
Encompass
Human IgG1 Poly-Specificity Control Antibody is a recombinant antibody that recognizes Baculovirus (BV) particles. Non-specific binding of an antibody to Baculovirus particles is a measure of the poly-specificity of that antibody.  H1308 is developed to be used as a control for poly-specific binding in analytical and molecular applications. For more information, please visit mednabio.com.
Catalog No. NC1907715