Learn More
Medchemexpress LLC HY-13646 100mg Medchemexpress, HM30181 HM30181A CAS:849675-66-7 Purity:98%
SDP
Catalog No. 501638005
Encompass_Preferred
Medchemexpress,HY-13646 100mg HM30181 HM30181A CAS:849675-66-7 Formula:C38H36N6O7 HM30181 is a potent and selective inhibitor of P-glycoprotein. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8005 Supplier Medchemexpress LLC Supplier No. HY13646100MG
Medchemexpress,HY-13646 100mg HM30181 HM30181A CAS:849675-66-7 Formula:C38H36N6O7 HM30181 is a potent and selective inhibitor of P-glycoprotein. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.