Learn More
Cayman Chemical HMMA (hydrochloride), Purity: ≥98%, CAS: 438625-58-2, 50 mg
SDP
Catalog No. 502954386
Encompass_Preferred
An analytical reference standard categorized as an amphetamine; an active metabolite of 3,4-MDMA; intended for research and forensic applications
Catalog No. 50-295-4386 Supplier Cayman Chemical Supplier No. 1419550 MG
An analytical reference standard categorized as an amphetamine; an active metabolite of 3,4-MDMA; intended for research and forensic applications