Learn More
TARGETMOL CHEMICALS INC HMAO-B-IN-4 5MG
SDP
Catalog No. 502579259
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hMAO-B-IN-4 is a selective reversible and blood–brain barrier (BBB) penetrable inhibitor of human monoamine oxidase-B (hMAO-B IC50=0.067 uM Ki= 0.03 uM hMAO-A IC50= 33.82 uM). purity: 99%
Catalog No. 50-257-9259 Supplier TARGETMOL CHEMICALS INC Supplier No. T677645MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hMAO-B-IN-4 is a selective reversible and blood–brain barrier (BBB) penetrable inhibitor of human monoamine oxidase-B (hMAO-B IC50=0.067 uM Ki= 0.03 uM hMAO-A IC50= 33.82 uM). purity: 99%