Learn More
Biocare Medical, LLC HMB45+MART-1+TYROSINASE 1.0ML
SDP
Catalog No. 5082576
Encompass_Preferred
HMB45 + MART-1 + Tyrosinase is a first-order pan melanoma screener. HMB45 labels the majority of melanomas. MART-1 is specific to melanocytic lesions. Tyrosinase labels a high percentage of desmoplastic melanomas.
Catalog No. 50-825-76 Supplier Biocare Medical, LLC Supplier No. CM165C
Add to Cart
Add to Cart
HMB45 + MART-1 + Tyrosinase is a first-order pan melanoma screener. HMB45 labels the majority of melanomas. MART-1 is specific to melanocytic lesions. Tyrosinase labels a high percentage of desmoplastic melanomas.