Learn More
Sigma Aldrich Fine Chemicals Biosciences HMBA AM resin NovabiochemR
Catalog No. 502908671
Encompass
This support consists of aminomethyl polystyrene resin functionalized with Sheppards base-labile 4-hydroxymethylbenzoic acid linker [1].The product number for this product was previously 01-64-0122.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-8671 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8550180025
This support consists of aminomethyl polystyrene resin functionalized with Sheppards base-labile 4-hydroxymethylbenzoic acid linker [1].The product number for this product was previously 01-64-0122.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.