Learn More
Aviva Systems Biology Corp HMBOX1 Antibody - N-terminal region (ARP39599_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39599_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HMBOX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HMBOX1. Alternate names: PBHNF, HNF1LAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53143