Learn More
Med Vet International Jorvet Cage Mounted Humidifier, with Flowmeter, J0953
Catalog No. 5000495219
Encompass_Preferred
The Jorvet Cage mounted oxygen delivery system, has a flowmeter with humidifer, and will atach to any cage. 15LPM flowmeter Bubble humidifer (J0653) add water or saline Attaches to cage bars via two U-bolts or wide flat washers Choices of male DISS fitting or male Ohio quick release for oxygen hose connection to flowmeter Oxygen tubing 7ft (j0656) for nasal insertiona or blow by
Catalog No. 50-004-95219 Supplier Med Vet International Supplier No. JOR953
The Jorvet Cage mounted oxygen delivery system, has a flowmeter with humidifer, and will atach to any cage. 15LPM flowmeter Bubble humidifer (J0653) add water or saline Attaches to cage bars via two U-bolts or wide flat washers Choices of male DISS fitting or male Ohio quick release for oxygen hose connection to flowmeter Oxygen tubing 7ft (j0656) for nasal insertiona or blow by