Learn More
Med Vet International Jorvet Cage Mounted Humidifier, with Flowmeter and Quick Connect, J0953Q
Catalog No. 5000495250
Encompass_Preferred
Jorvet's cage mounted oxygen deliver system with has a flowmeter with humidifier. This oxygen delivery system can attach to any cage. 15LPM flowmeter Bubble humidifier, add water or saline Attaches to cage bars via two U bolts or wide flat washers Choice of male DISS fitting or male Ohio quick release for oxygen hose connection to flowmeter 7ft. Oxygen Tubing 14fr oxygen catheter for nasal insertion ro blow by
Catalog No. 50-004-95250 Supplier Med Vet International Supplier No. JOR953Q
Jorvet's cage mounted oxygen deliver system with has a flowmeter with humidifier. This oxygen delivery system can attach to any cage. 15LPM flowmeter Bubble humidifier, add water or saline Attaches to cage bars via two U bolts or wide flat washers Choice of male DISS fitting or male Ohio quick release for oxygen hose connection to flowmeter 7ft. Oxygen Tubing 14fr oxygen catheter for nasal insertion ro blow by