Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMDI, MilliporeSigma™ Supelco™

Catalog No. 111019740 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9740 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9740 Supplier MilliporeSigma Supelco Supplier No. 334902MLR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 5124-30-1
Flash Point 200°C - Closed Cup
Physical Form Neat
Linear Formula CH2(C6H10 NCO)2
MDL Number MFCD00015405
Synonym 4,4 ′-Methylene-bis(cyclohexyl isocyanate); 4,4 ′-Diisocyanato-methylenedicyclohexane; 4,4 ′-HMDI; Bis(4-isocyanatocyclohexyl)methane
Type Analytical Standard
Quantity 2 mL
Formula Weight 262.35
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.