Learn More
Lonza Walkersville HMEC - Hu Mammary Epit Cells
Catalog No. 5000496361
Encompass
Lonza s Human Mammary Epithelial Cells (HMEC) are isolated from adult female breast tissue The cells have been primarily utilized to understand the formation of breast cancer Cryopreserved Human Mammary Epithelial Cells are guaranteed through 15 population doublings express cytokeratins 14 and 18 and are negative for cytokeratin 19
Catalog No. 50-004-96361 Supplier Lonza Walkersville Supplier No. CC2551
Lonza s Human Mammary Epithelial Cells (HMEC) are isolated from adult female breast tissue The cells have been primarily utilized to understand the formation of breast cancer Cryopreserved Human Mammary Epithelial Cells are guaranteed through 15 population doublings express cytokeratins 14 and 18 and are negative for cytokeratin 19