Learn More
Active Motif hMeDIP
SDP
Catalog No. NC1852697
Encompass
The hMeDIP Kit is designed to immunoprecipitate and enrich for DNA fragments containing 5-hydroxymethylcytosine (5-hmC). The hMeDIP Kit contains a highly specific purified 5-hmC antibody and the necessary buffers to perform methylated DNA immunoprecipitation (MeDIP). The affinity of the antibody enables separation of 5-hmC DNA from 5-mC DNA. Additionally, the antibody works to efficiently immunoprecipitate either double-stranded or single-stranded input DNA.
Catalog No. NC1852697 Supplier Active Motif Supplier No. 55010
The hMeDIP Kit is designed to immunoprecipitate and enrich for DNA fragments containing 5-hydroxymethylcytosine (5-hmC). The hMeDIP Kit contains a highly specific purified 5-hmC antibody and the necessary buffers to perform methylated DNA immunoprecipitation (MeDIP). The affinity of the antibody enables separation of 5-hmC DNA from 5-mC DNA. Additionally, the antibody works to efficiently immunoprecipitate either double-stranded or single-stranded input DNA.