Learn More
Cayman Chemical HMG-CoA Reductase human reco
SDP
Catalog No. 502969974
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 76.5 kDa
Catalog No. 50-296-9974 Supplier Cayman Chemical Supplier No. 14944250 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 76.5 kDa