Learn More
Aviva Systems Biology Corp HMG20A Antibody - N-terminal region (ARP39339_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39339_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMG20A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HMG20A. Alternate names: FLJ10739, HMGX1, HMGXB1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-56990