Learn More
Enzo Life Sciences HMGB 1 monoclonal antibody (KS1) (100µg)
SDP
Supplier:  Enzo Life Sciences ADICSA614E
Encompass_Preferred
Clone: KS1. Host: Mouse. Isotype: IgG2a. Immunogen: Pig HMGB1 (HMG1). Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7192