Learn More
Aviva Systems Biology Corp HMGB1 Antibody - middle region (ARP35646_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35646_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HMGB1. Alternate names: DKFZp686A04236, HMG1, HMG3, SBP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55803