Learn More
Aviva Systems Biology Corp HMGB1 Antibody - N-terminal region (ARP59013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HMGB1. Alternate names: DKFZp686A04236, HMG1, HMG3, SBP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-557