Learn More
Chondrex Inc HMGB1 ELISA, 1 KT
Supplier:  Chondrex Inc 6010
Encompass_Preferred
ELISA kit for determining HMGB1 levels in cell culture media, serum, and plasma samples. Reactivity: Mouse, Rat, Bovine, Human. Standard Range: 50 ng/mL to 0.78 ng/mL. Working Time: 2 hours (2-day assay).
Catalog No. 50-152-6779