Learn More
ACROBiosystems ACROBiosystems Mouse HMGB1 Protein, His Tag
Supplier:  ACROBiosystems HM1M52H6100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Glu 215 (Accession # P63158-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.8 kDa. The protein migrates as 33 kDa and 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6885