Learn More
ACROBiosystems Recombinant Protein;Human HMGB1 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;HM1,HMGB1,HMG1,HMG3,SBP-1,HMGB-1,HMG-1,HMG-3,SBP1
Catalog No. 501657919
Encompass_Preferred
Recombinant Protein;Human HMGB1 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human HMGB1, Mouse IgG2a Fc Tag, low endotoxin (HM1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Glu 215 (Accession # P09429-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 51.6 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7919 Supplier ACROBiosystems Supplier No. HM1H5255100UG
Recombinant Protein;Human HMGB1 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human HMGB1, Mouse IgG2a Fc Tag, low endotoxin (HM1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 2 - Glu 215 (Accession # P09429-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 51.6 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.