Learn More
Aviva Systems Biology Corp HMGB2 Antibody - middle region (ARP58158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HMGB2. Alternate names: HMG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62252