Learn More
Aviva Systems Biology Corp HMGB3 Antibody - C-terminal region (ARP35765_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35765_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HMGB3. Alternate names: HMG4, HMG-4, HMG2A, HMG-2aApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55841