Learn More
Aviva Systems Biology Corp HMGCLL1 Antibody - N-terminal region (ARP56277_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56277_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGCLL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HMGCLL1. Alternate names: DKFZP434G1411, bA418P12.1ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61793