Learn More
Aviva Systems Biology Corp HMGCS2 Antibody - C-terminal region (ARP41563_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41563_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGCS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HMGCS2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57907