Learn More
Aviva Systems Biology Corp HMGN1 Antibody - middle region (ARP38532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMGN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HMGN1. Alternate names: FLJ27265, FLJ31471, HMG14, MGC104230, MGC117425ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56691