Learn More
Aviva Systems Biology Corp Hmgn2 Antibody - middle region (ARP51631_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51631_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hmgn2. It was validated on Western Blot. . Alternate names: HMG-17, Hmg17, MGC102472, MGC103137, MGC103384, MGC118420ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-581