Learn More
Electron Microscopy Sciences BeadBug™ Homogenizer Beads in Prefilled Tube, Standard Glass 0.5 mm
SDP
Catalog No. 501931126
Encompass_Preferred
Acid washed glass beads for BeadBug microtube homogenizer.
Includes: Standard Glass Beads
Diameter: 0.5 mm
Catalog No. 50-193-1126 Supplier Electron Microscopy Sciences Supplier No. 7262004
Acid washed glass beads for BeadBug microtube homogenizer.
Includes: Standard Glass Beads
Diameter: 0.5 mm