Learn More
Electron Microscopy Sciences BeadBug™ Homogenizer Beads in Prefilled Tube, High Impact Zirconium Assorted sizes
SDP
Catalog No. 501931133
Encompass_Preferred
Triple-Pure molecular biology grade beads for BeadBug microtube homogenizer.
Includes: High Impact Zirconium Beads
Diameter: 0.1, 0.5, 1.0, 1.5, 3.0 mm
Catalog No. 50-193-1133 Supplier Electron Microscopy Sciences Supplier No. 7262011
Triple-Pure molecular biology grade beads for BeadBug microtube homogenizer.
Includes: High Impact Zirconium Beads
Diameter: 0.1, 0.5, 1.0, 1.5, 3.0 mm