Learn More
Electron Microscopy Sciences BeadBug™ Homogenizer Beads in Prefilled Tube Zirconium 0.1 mm
SDP
Catalog No. 501931128
Encompass_Preferred
Triple-Pure molecular biology grade beads for BeadBug microtube homogenizer.
Includes: Zirconium Beads
Diameter: 0.1 mm
Catalog No. 50-193-1128 Supplier Electron Microscopy Sciences Supplier No. 7262006
Triple-Pure molecular biology grade beads for BeadBug microtube homogenizer.
Includes: Zirconium Beads
Diameter: 0.1 mm