Learn More
Grainger Brady GHS Label, Vinyl, Adhesive, 3 in x 5 in, 50/Pk.
Catalog No. 502761242
Encompass_Preferred
These GHS-compliant right-to-know labels feature a durable, flexible vinyl construction designed to adhere permanently to rough, textured, and challenging surfaces. Utilizing a permanent acrylic adhesive, these labels are engineered for robust performance in both indoor and outdoor environments, offering resistance to moisture and chemicals.
  • Permanent acrylic adhesive ensures reliable adhesion to a wide variety of surfaces
  • Constructed from flexible vinyl film (B-7569) for durability
  • Withstands exposure to moisture, chemicals, and handling
  • Supports a service temperature range of -4°F to 167°F (-20°C to 75°C)
  • Features an average outdoor durability of up to 3 years
  • Includes a write-on surface for additional documentation
Catalog No. 50-276-1242 Supplier Grainger Supplier No. 15Y474
These GHS-compliant right-to-know labels feature a durable, flexible vinyl construction designed to adhere permanently to rough, textured, and challenging surfaces. Utilizing a permanent acrylic adhesive, these labels are engineered for robust performance in both indoor and outdoor environments, offering resistance to moisture and chemicals.
  • Permanent acrylic adhesive ensures reliable adhesion to a wide variety of surfaces
  • Constructed from flexible vinyl film (B-7569) for durability
  • Withstands exposure to moisture, chemicals, and handling
  • Supports a service temperature range of -4°F to 167°F (-20°C to 75°C)
  • Features an average outdoor durability of up to 3 years
  • Includes a write-on surface for additional documentation