Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ HMIG Labels Station

Catalog No. 19112820 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Brady Label Printers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-112-820 Brady Label Printers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-112-820 Supplier Brady™ Supplier No. PD674E
Add to Cart
Add to Cart

HMIG Labels Station provides employees with hazard rating information and keeps HMIG labels organized.

  • Six 3/4 in. pockets for holding sheet labels
  • One 5 in. dowel to hold label rolls
  • Includes a pen holder
  • Provides hazard rating informationprinted on board

Specifications

Product Type HMIG Labels Station
For Use With (Equipment) Brady Label Printers
Dimensions (L x W) 11 x 18 in.
Sterility Non-sterile
Includes Six 0.75 in. pockets for holding sheet labels and one 5 in. dowel to hold label rolls and pen holder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.